Key Documents
SF-UC-500-100
Novas Bio
Novas Bio Sealing Film
100 Films - Ultra clear, optical Sealing Film with pressure-sensitive adhesive. For qPCR.
Synonym(s):
PCR sealing film, qPCR, real-time PCR
About This Item
PDF Icon
Shipped & Sold By Novas Bio
A Trusted Marketplace Seller
PDF Icon
Have a question?
material
polyolefin
W × L
3 in x 5.5 in
General description
  • Highest optical clarity and low auto-fluorescence for real-time PCR
  • Pressure-sensitive silicone adhesive is medical-grade, non-absorbing, and non-fluorescing
  • The non-tacky adhesive simplifies film handling and positioning, and minimizes contamination
  • Easy-peel, perforated plastic liner
  • Chemically resistant to DMSO, acetonitrile, and methanol
  • Universal cut film for use with a variety of plate formats, including flat-top, non-skirted, raised-rim and semi-skirted PCR plates
  • Thickness: 147 μm / 5.8 mil
  • Temperature Range: -80 °C to +105 °C
  • Certified DNase/RNase/Human DNA free. Non-pyrogenic.
Choose from one of the most recent versions:
Certificates of Analysis (COA)