Key Documents
FT-200-R-S-10
Novas Bio
Novas Bio Filter Pipette Tips
96 Tips/Rack, 10 Racks per Unit - 200 uL Filtered Tip, Sterile
Synonym(s):
Filter tip, pipette tip
About This Item
PDF Icon
Shipped & Sold By Novas Bio
A Trusted Marketplace Seller
PDF Icon
Have a question?
binding type
non-treated surface
feature
graduations: Yes
material
polypropylene
volume
0.2 mL
200 μL
General description
  • Racked in 96-well format. 96 tips/rack x 10 racks/unit.
  • Engineered to fit most universal single and multi-channel pipettors
  • Manufactured with medical-grade polypropylene
  • Certified DNase/RNase/Human-DNA free. Non-pyrogenic
  • Exceptional clarity for maximum sample visibility
  • Filtered tip prevents aerosol and liquid contamination
  • Pre-sterilized by E-beam irradiation
Legal Information
3926.90.9910
Choose from one of the most recent versions:
Certificates of Analysis (COA)