175CM-100MLX
Monobind Inc.
Monobind Inc. Human Serum Lipid Stripped
Mixed Gender, Pooled - 100 ml
Synonym(s):
Human Serum, Control, Calibrator, Standard, LC-MS, MS, GCMS, LCMS, Mass Spectrometry, Assays
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
form
liquid
storage temp.
< 8 °C
storage condition
avoid repeated freeze/thaw cycles
General description
Human Serum - Mixed Gender - produced by removing clotting factors and lipid stripping (removing triglycerides and cholesterol) to obtain a stable matrix. For use in various professional-lab applications such as preparing controls, calibrators, and standards for assays and LC-MS/MS technology.
- Shelf Life: 5 years from DOM at 8°C or Colder.
- Packaging & Shipping: 100ml in Media Bottle. Ships Frozen with Cool Packs, Overnight Only.
- Filtration: 0.2um absolute membrane
- Characteristics: Total Protein, 5.5 to 6.0 grams/ dL. A Certificates of Analysis (COA) will confirm that this product has met all performance specifications prior to release for sale and accompanies each lot.
- Origin & Testing*: US Collection Centers - Donor Testing negative for (HIV-1&2, HCV, HBsAg, HIV-Ag, ALT and RPR) by FDA Method. *May vary by lot, refer to the provided COA.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.