MilliporeSigma
All Photos(4)
Key Documents
403003012-220
Lighthouse Worldwide Solutions
LWS SHIELD-95-PRO N95 Respirator Mask
N95 Respirator Mask
Synonym(s):
N95 Respirators, Protective Face Masks, NIOSH-approved N95, Airborne Particle Protection, Respiratory Protection, Personal Protective Equipment, PPE, Surgical Mask Alternatives, Virus Filtration, Efficiency SHIELD-PRO N95 Masks
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Lighthouse Worldwide Solutions
General description
LWS has developed the SHIELD N95 disposable respirator to fit more comfortably and securely than any other respirator on the market. The SHIELD focuses on what matters most to the end-user: fit, breathability, visibility, and durability.
  • Designed for construction, first responders, and general use for those not requiring surgical rated N95 respirators
  • 92% Fit test passing on a wide range of faces
  • Particle filtration: greater than 98%
  • Designed to make breathing easier and reduce fogging
  • Improved ergonomics and expanded peripheral visibility
  • Maximimum durability
  • Individually wrapped packaging
  • Made in the USA
The SHIELD N95 respirator has been declared the 'fit test champion' by many users and fit testing consultants. The innovative flat-fold, 3-panel design and soft nose foam allows for a more comfortable and secure fit on a wide range of face shapes and sizes. The SHIELD is one of the most breathable disposable masks on the market, with 15% more breathability than the leading N95 mask. This respirator's breathable design, coupled with its specialized closed-cell nose foam and secure fit, helps to alleviate the annoying problem of eyewear fogging.
N95 masks can obstruct a portion of the lower peripheral vision, forcing wearers to tilt their head forward and increasing strain and fatigue on the neck. The SHIELD respirator has a patent pending 'ClearView Bar' feature that helps reduce neck strain by removing the vision blockage that can occur when wearing an N95 mask.
The SHIELD-95-PRO N95 respirator features a polypropylene filter, medical grade PVC nose foam, a dual Iron Core polypropylene-coated wire nose clip, polypropylene coverweb, and polyisoprene head straps.
Shelf/storage life for this item is 3 years. Respirators should be stored in temperatures between −4 to 86 °F (−20 to 30 °C), not exceeding 80% relative humidity.
Certifications: NIOSH # TC 84A-9466. ASTM F2101. 16 CFR 1610. 42 CFR 84.
Ordering information: Respirators are packaged 20 per carton, 4 cartons per case (80 total per case). Minimum order quantity: 1 case.
Caution: N95 respirators reduce exposure to many forms of airborne particulates. Read User Instructions in the packaging. Users must comply with OSHA 1910.134 for program, training, fit testing, and evaluation.