Key Documents
HS3902
Hausser Scientific
Hausser Scientific™ Petroff Hausser Bacteria Counter
Counting chamber with cover glass and two grid, 20 microns
Synonym(s):
hemacytometer, counting chamber
About This Item
PDF Icon
Shipped & Sold By HAUSSER SCIENTIFIC CO.
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Hausser Scientific
General description
The Petroff-Hausser Counter is one piece construction ensuring durability and accuracy, featuring an Improved Neubauer ruling on a single plateau. The Petroff-Hausser Counting Chamber is popular for bacteria and sperm counting and offered in a series of cell-depths. This chamber is 20 microns deep with two counting areas.
Choose from one of the most recent versions:
Certificates of Analysis (COA)