099A-PPM12
Glas-Col®
Glas-Col® Digital Pulse Mixer
Pulse Mixer
Synonym(s):
Mixing Sample Prep
About This Item
manufacturer/tradename
Glas-Col
General description
Digital Pulse Mixer base.
Glas-Col's new orbital Pulse Mixer is capable of performing high or low speed pulse mixing for most applications. This unit is able to accommodate a wide variety of glassware, racks, well plates or reactor blocks for accurate, repeatable results.
Our pulse mixers have been used in blood banks, clinical labs, water testing facilities (EPA methods) and many other research labs.
In the QuEChERS procedure, our systems are well adapted for the various vials associated with this method and can mix multiple samples for high throughput testing.
- 100-1500rpm
- Digital readout
- 120 Volts
- Size 16"w x 18"d x 17" H
- Platform size 11-5/8” x 14-1/4"
- Mixer mounts Vibration absorbing
- On/Off switch Located on front of unit
- Finish Silver and black powder coat
- Cord 3-wire 6' long
- Load Up to 10 lbs.