Key Documents
BI0139
Eisco Labs
Eisco Labs 8 Piece Plastic Sieve/ Strainer
8 Piece Plastic Sieve/ Strainer
Synonym(s):
Sieve, Strain, Sift, Filter
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • Six plastic sieves with stainless steel mesh, cover and Bottom Collector.
  • Each Sieve has a plastic frame having an individual size one each ( Mesh no. 5, 10, 35, 60, 120, 230)
  • Complete with plastic lid and bottom collector
  • Stackable - Diameter of Sieves are 6.75"
  • Total height of stacked sieves approx. 15"
Eisco Labs 8 Piece Plastic Sieve/ Strainer - Stainless Steel Mesh Six plastic sieves with stainless steel mesh. Each Sieve has a plastic frame having an individual size one each ( Mesh no. 5, 10, 35, 60, 120, 230) Complete with plastic lid and bottom collector. Stackable, Total height of stacked sieves approx. 15.25" Not not food.
Choose from one of the most recent versions:
Certificates of Analysis (COA)