9025
CELLTREAT
VistaLab Pipette Tips
250µL Pipette Tips, MLA, Stacked Dense Rack, Non-sterile
Synonym(s):
pipette tips
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
CELLTREAT
General description
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Stacked Rack- Non-sterile only
- Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved
- 200 tips/layer
- For Small volume (250 mL) tips - 5 layers - 1000 tips
- For Large volume (1250 mL) tips - 3 layers - 600 tips