CW5601
Alkali Scientific™
Alkali Scientific™ CellPro™ 15mL MARKER Conical Tubes
15mL , Centrifuge Tubes, Clear Polypropylene, Conical bottom w/ WHITE Screw Caps, Resealable bags, 500/Case
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Alkali Scientific™
General description
CellPro™ 15mL MARKER™ Conical Centrifuge tubes are conical bottom, tubes.
The unique MARKER top allows for easy identification of sample on the top. This application is especially suited for making with a SHARPIE or similar permanent marker. The non-smearing surface on the top of the cap maintains sample ID even after multiple freeze / thaw cycles.
MARKER™ tubes have an industry leading, largest available white writing surface, commonly 35% larger than other similar tubes. Crisp, legible graduations in black print allow contrast for quick volume reference. Ultra clear polypropylene provides unmatched optical optical clarity. Improved your sample visibility.
MARKER™ tubes are available racked configuration or bagged configuration. Green Lab Certified, outer cartons are recyclable, inner racks are recyclable and inner bags are recyclable. Each inner sealed pack is sterilized for aseptic distribution within your lab or sample collection site.
MARKER™ Tubes are non-pyrogenic and free of RNase, DNase, and human DNA. Entire contents are non-cytotoxic MARKER™ Tubes are highly resistant to any organic solvents and exhibit a stable sample collection & processing environment for nearly all liquids and solids.
Legal Information
CellPro™ is a trademark of Alkali Scientific.
Choose from one of the most recent versions:
Certificates of Analysis (COA)