MilliporeSigma
All Photos(1)
Key Documents
B6916
Supelco
Bradford Reagent
for 0.1-1.4 mg/ml protein
Synonym(s):
Coomassie dye binding protein assay, Coomassie dye binding protein assay, Protein dye reagent, Protein dye reagent
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12161500
NACRES:
NA.32
Quality Level
form
solution
storage temp.
2-8°C
General description
Bradford assay is addition of coomassie brilliant blue G-250 to protein solution. The coomassie blue dye associates with basic and aromatic amino acids, thereby causing shift in absorbance during protein determination.
Application
Bradford Reagent has been used to determine total protein concentration.
Features and Benefits
  • The reagent is ready to use. No mixing or dilution required.
  • Color development is rapid. Only a five minute incubation and then the sample is read a 595 nm.
  • Reducing sugars and reducing substances along with thiols do not interfere with this reagent.
  • Reagent is suitable for micro (1-10 μg/ml) and standard (50-1400 μg/ml) assays.
  • Can be used in microwell plate assays.
  • Inexpensive assay.
Legal Information
Application
Product No.
Description
Pricing
also commonly purchased with this product
related product
Product No.
Description
Pricing
pictograms
Health hazardCorrosion
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 2
target_organs
Eyes,Central nervous system
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Role of rpoS in the development of cell envelope resilience and pressure resistance in stationary-phase Escherichia coli.
Charoenwong D et al.
Applied and Environmental Microbiology, 77, 5220-5229 (2011)
Sin-Jin Li et al.
Clinical nutrition (Edinburgh, Scotland), 36(3), 760-767 (2016-06-28)
The cellular mechanisms of obesity-induced cardiomyopathy are multiple and not completely elucidated. The objective of this study was to differentiate two obesity-associated cardiomyopathy miniature pig models: one with the metabolic syndrome (MetS), and one with a metabolically healthy obesity (MHO).
M Tal et al.
The Journal of biological chemistry, 260(18), 9976-9980 (1985-08-25)
Dimethyl sulfoxide was found to be effective for extraction of Coomassie Brilliant Blue R-250 (Coomassie R) from stained proteins on polyacrylamide gel slices. A good correlation was found between the ability of different proteins to bind Coomassie R and their
Influence of condensing equipment and temperature on exhaled breath condensate pH, total protein and leukotriene concentrations.
Czebe K et al.
Respiratory Medicine, 102, 720-720 (2008)
Kapil Dev Singh et al.
PloS one, 11(3), e0149418-e0149418 (2016-03-18)
Complex traits, including common disease-related traits, are affected by many different genes that function in multiple pathways and networks. The apoptosis, MAPK, Notch, and Wnt signalling pathways play important roles in development and disease progression. At the moment we have
Articles
The field of proteomics is continually looking for new ways to investigate protein dynamics within complex biological samples. Recently, many researchers have begun to use RNA interference (RNAi) as a method of manipulating protein levels within their samples, but the ability to accurately determine these protein amounts remains a challenge. Fortunately, over the past decade, the field of proteomics has witnessed significant advances in the area of mass spectrometry. These advances, both in instrumentation and methodology, are providing researchers with sensitive assays for both identification and quantification of proteins within complex samples. This discussion will highlight some of these methodologies, namely the use of Multiple Reaction Monitoring (MRM) and Protein-AQUA.
Protocols
To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Related Content
Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.
Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service