MilliporeSigma
Intensified and Continuous Downstream Processing
Gain operational efficiency and flexibility while reducing footprint and costs
HomeIntegrated OfferingsBiopharma 4.0Intensified & Continuous Downstream ProcessingFlow-Through Polishing Platform
Looking to increase your downstream process efficiency?
Transformation of the biomanufacturing workflow is being accelerated by the demand for new therapeutics combined with the need to reduce costs while ensuring operational flexibility and productivity.
Process intensification, closed, and continuous bioprocessing are foundational strategies for achieving these goals. They are widely and successfully deployed in the downstream suite to be more efficient, faster, smaller, less expensive, and sustainable.
To help biopharmaceutical manufacturers intensify, closing, digitally-enhance, or go fully continuous with their processes and gain new levels of operational efficiency and flexibility, there are multiple levers within the DSP suite.
Flow-Through Polishing Chromatography
Polishing chromatography occurs post viral inactivation of mAb downstream processing to remove any remaining impurities from the monoclonal antibody feed stream. In traditional batch processes, polishing steps comprise nearly 25% of the cost of production. Integrating individual polishing steps into a single, continuous flow-through process delivers increased efficiency and yields while reducing costs by up to 10%. This intensified approach lowers the overall footprint through the elimination of intermediate hold tanks, buffer consumption reduction, and the removal of product dilution steps.  Overhead costs such as operator intervention are further reduced by the integration of process control systems.
  •  Eshmuno® CP-FT Resin, designed to provide efficient removal of mAb aggregates in the flow-through frontal chromatography mode of operation enabling loading capacities 10× higher than traditional bind/elute CEX chromatography.
  • Natrix® Q Chromatography Membrane, a high capacity, high throughput strong anion exchange membrane adsorber designed to purify biomolecules in flow-through operating conditions.
Related Categories
Operator, fully gowned, standing in front of the Mobius Multi Column Capture System
Chromatography Systems for Bioprocessing
Single-use and multi-use chromatography systems are critical for the separation of your valuable molecules.
4 different sizes of Affinity chromatography with Eshmuno® resin bottles sit on a black counter with a scientist working on a piece of machinery in the background
Affinity Chromatography Resins
Affinity chromatography is often used during the capture phase to isolate the target molecule and reduce process volume.
4 different sizes of Eshmuno CMX, ion exchange chromatography resin bottles
Ion Exchange Chromatography Resins
Ion exchange (IEX) chromatography separates biomolecules based on difference in charge between the target molecule and chromatography resin.
3 sizes of Natrix Q chromatography membrane devices
Membrane Chromatography
Optimize downstream purification using membrane chromatography for capture and polishing. Natrix membranes boost economics and facility efficiency.
Pelicon cassette lab-scale single-pass tangential flow filtration setup
Tangential Flow Filtration Devices
Elevate Filtration Precision: Our tangential flow products ensure purity for mAbs, vaccines, and more, scaling from single-use to multi-use.
Related Resources
Related Webinars
Effective Implementation of Multi Column Chromatography
Improve your process efficiency and reduce costs
Related Videos
BioContinuum™ Platform: Your enabler of the biomanufacturing facility of the future!