MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Plasmid DNA | Plasmid Purification Kits
Circular plasmid DNA strand, showcasing the double helix structure with alternating colors representing the base pairs.
Plasmid purification is commonly used for molecular cloning, transformation, and recombinant protein expression. Our plasmid purification kit essentials are available based on the size of the bacterial culture and corresponding plasmid yield (miniprep, midiprep, maxiprep, megaprep, and gigaprep). Our plasmid kits provide reliable isolation of high-quality plasmid DNA. These kits are designed with advanced silica bind and elute technology. Additionally, our kits are compatible with both vacuum and spin formats and formulated without phenol/chloroform.
For incremental innovations, day after day, choose plasmid DNA purification kits and essentials that are the foundation of great work, not a variable that undermines it. Our plasmid purification kits allow researchers to maximize the reproducibility of their workflow and minimize the time spent obtaining plasmid DNA for transfection, sequencing, PCR, and other downstream applications. Explore all of our plasmid DNA purification kits below.
Products
kits (14)
plates (1)
Roche (2)
Millipore (2)
Sigma-Aldrich (4)
Sort by Relevance
Showing 1-15 of 15
Change View
Showing 1-15 of 15
GenElute<SUP>™</SUP> Plasmid Miniprep Kit
PLN70
GenElute Plasmid Miniprep Kit
sufficient for 70 purifications
GenElute<SUP>™</SUP> Plasmid Miniprep Kit
PLN350
GenElute Plasmid Miniprep Kit
sufficient for 350 purifications
Genopure Plasmid Midi Kit
03143414001
Genopure Plasmid Midi Kit
kit of for 20 isolations from 5 to 30 ml
Genopure Plasmid Maxi Kit
03143422001
Genopure Plasmid Maxi Kit
kit of for 10 isolations from 30 to 150 ml
GenElute<SUP>™</SUP> HP Select Plasmid Gigaprep Kit
NA0800
GenElute HP Select Plasmid Gigaprep Kit
sufficient for 5 preparations
GenElute<SUP>™</SUP> Endotoxin-free Plasmid Midiprep Kit
PLED35
GenElute Endotoxin-free Plasmid Midiprep Kit
sufficient for 35 purifications
GenElute<SUP>™</SUP> Plasmid Midiprep Kit
PLD35
GenElute Plasmid Midiprep Kit
sufficient for 35 purifications
Plasmid Miniprep<sub>96</sub> Kit
LSKP096
Plasmid Miniprep96 Kit
Plasmid & BAC Miniprep
GenElute<SUP>™</SUP> HP Plasmid Miniprep Kit
NA0150
GenElute HP Plasmid Miniprep Kit
sufficient for 70 preparations
GenElute<SUP>™</SUP> HP Plasmid Miniprep Kit
NA0160
GenElute HP Plasmid Miniprep Kit
sufficient for 350 preparations
GenElute<SUP>™</SUP> HP 96-Well Plasmid Miniprep Kit
NA9604
GenElute HP 96-Well Plasmid Miniprep Kit
sufficient for 4 96-well plate purifications
GenElute<SUP>™</SUP> Endotoxin-free Plasmid Maxiprep Kit
PLEX15
GenElute Endotoxin-free Plasmid Maxiprep Kit
sufficient for 15 purifications
GenElute<SUP>™</SUP> Plasmid Maxiprep Kit
PLX15
GenElute Plasmid Maxiprep Kit
sufficient for 15 purifications
GenElute<SUP>™</SUP> HP Endotoxin-Free Plasmid Maxiprep Kit
NA0400
GenElute HP Endotoxin-Free Plasmid Maxiprep Kit
sufficient for 10 preparations
Multiscreen HTS Filter Plate, Styrene
MSNANLY
Multiscreen HTS Filter Plate, Styrene
Clearing of bacterial cell lysates
You have viewed 1-15 of 15 results
Page 1 of 1
Page 1 of 1
GenElute™ plasmid miniprep kits
GenElute™ plasmid miniprep kits offer simple, rapid, and cost-effective methods for isolating plasmid DNA from E. coli cultures. These kits combine silica-based membrane technology and the convenience of a spin column format.
Features:
  • 40% more preps per kit than the leading supplier
  • Purify up to 20 µg of plasmid DNA per mL of culture
  • Purified plasmid DNA in less than 30 minutes for up to 24 preps
  • No phenol/chloroform extraction or alcohol precipitation required
GenElute™ HP plasmid prep kits
GenElute™ HP plasmid purification kits yield high-quality plasmid DNA in less than 30 minutes for Mini, Midi, and Maxiprep kits, or 1.5 hours for Mega and Gigaprep kits.
  • From harvested bacterial culture to pure plasmid DNA in less time
  • Up to 25 µg (Mini), 350 µg (Midi), 1.2 mg (Maxi), 5 mg (Mega), and 15 mg (Giga) yield of high-copy plasmid DNA
  • Offers the flexibility of a vacuum or spin format (Mega and Gigaprep in vacuum format only)
  • No phenol/chloroform extraction or alcohol precipitation required
  • Kits are stable at room temperature for convenient storage
GENOPURE™ PLASMID PREP KITS
Roche Genopure™ midi and maxi prep kits use anion exchange chromatography for plasmid purification. A modified alkaline lysis protocol optimized for cell cultures grown in LB media is used to isolate DNA. The lysate can be filtered rather than centrifuged prior to anion exchange, resulting in shorter plasmid prep times and complete removal of SDS from the purified product.
  • Reduce DNA shearing by clarifying lysate using pre-folded filters rather than centrifugation prior to anion exchange.
  • Save time with ready-to-use reagents (10 minutes hands-on time/75 minutes overall).
  • Purify all sizes and types of plasmid, even BAC DNA.
  • Process multiple samples in parallel using high-speed gravity-flow columns.
  • Eliminate the use of hazardous organic compounds such as cesium chloride, phenol, chloroform, and ethidium bromide.
GenElute™ endotoxin-free plasmid prep kits
Endotoxins reduce cell viability and transfection efficiencies among different eukaryotic cell lines and can influence the outcome and reproducibility of a given transfection experiment when using mammalian and other eukaryotic cells. GenElute™ endotoxin-free plasmid prep kits offer a simple, rapid, cost-effective method for purifying plasmid DNA with <0.1
EU/mg DNA for high-efficiency transfection.
  • Higher transfection efficiency and yield than market leaders
  • Purify endotoxin-free plasmid DNA (≤0.1 EU/mg DNA)
GenElute™ five-minute plasmid miniprep kits
GenElute™ five-minute miniprep kits feature a streamlined protocol yielding up to 5 mg high-quality plasmid DNA in about five minutes.
  • Only 5 minutes from start to finish
  • No pelleting cells or clearing lysates
  • Binding column works with any standard laboratory vacuum manifold
  • Choose vacuum or spin protocols
Related Resources
Page 1 of 2
Web Tools