Certificate of Analysis
Product Name:
Water
Product Description:
sterile-filtered, BioReagent, suitable for cell culture
Product Brand:
Sigma-Aldrich
Product Number:
W3500
Molecular Weight:
18.02
Molecular Formula:
H2O
CAS Number:
7732-18-5
Test
Specification
Lot RNBL9398 Results
Storage:
ROOM TEMPERATURE
Print Date:
06 MAY 2023
Expiry date:
APR 2025
Date of QC Release:
06 MAY 2023
Place of Manufacture:
Irvine. United Kingdom
Production Date:
APR 2023
Appearance (Turbidity)
Clear
Clear
Appearance (Form)
Liquid
Liquid
pH
5.0 - 7.0
5.7
Sterility
Pass
Pass
Endotoxin Level
<= 1 EU/ml
< 1 EU/ml
Cell Culture Testing - MTT
Pass
Pass
Cell Line
Cell Line - Cell Types
L929