MilliporeSigma
Certificate of Analysis
Product Name:
Monoclonal Anti-VGAT antibody produced in mouse
Product Description:
Prestige Antibodies® Powered by Atlas Antibodies, clone CL2793, purified immunoglobulin, buffered aqueous glycerol solution
Product Brand:
Sigma-Aldrich
Product Number:
AMAB91043
Test
Specification
Lot 3008 Results
Immunohistochemistry Dilution
1:50 - 1:200
PASS
Immunohistochemistry Antibody Retrieval Method
HIER pH6
PASS
Concentration
Report result
0.5µg/ml
quality signature