Tris base
Synonym(s):
Trizma® base, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol, Tris, Tromethamine
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
pK Value
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing