Skip to Content
MilliporeSigma
Trifloxystrobin PESTANAL®, analytical standard

Trifloxystrobin

Trifloxystrobin PESTANAL®, analytical standard
Synonym(s):
Methyl (E)-α-methoxyimino-2-[(E)-1-(3-trifluoromethylphenyl)ethylidenaminooxymethyl]phenylacetate
Empirical Formula (Hill Notation):
C20H19F3N2O4
CAS Number:
Molecular Weight:
408.37

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing