Sulfuric acid
Synonym(s):
Sulfuric acid, Sulfuric acid solution
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Compatibility
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-20 of 26
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2