Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium formate
Synonym(s):
Formic acid sodium salt, Sodium formate
Linear Formula:
HCOONa
CAS Number:
141-53-7
Molecular Weight:
68.01
EC Number:
205-488-0
Products
Shipping
Ships Today (6)
Product Category
salts (4)
Brand
Sigma-Aldrich (5)
Supelco (2)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
ACS reagent (2)
analytical standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
247596
Sodium formate,
ACS reagent, ≥99.0%
Expand
View Pricing
107603
Sodium formate,
reagent grade, 97%
Expand
View Pricing
71539
Sodium formate,
BioUltra, ≥99.0% (NT)
Expand
View Pricing
456020
Sodium formate,
99.998% trace metals basis
Expand
View Pricing
1.06443
Sodium formate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
798630
Sodium formate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
67253
Sodium formate,
analytical standard
Expand
View Pricing