Skip to Content
MilliporeSigma
Sodium fluoride JIS special grade, ≥99.0%

Sodium fluoride

Sodium fluoride JIS special grade, ≥99.0%
Synonym(s):
Sodium fluoride, Sodium Fluoride
Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique