Skip to Content
MilliporeSigma
Sodium dodecyl sulfate BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri™

Sodium dodecyl sulfate

Sodium dodecyl sulfate BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri™
Synonym(s):
Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate, Sodium dodecyl sulfate, Sodium n-Dodecyl Sulfate, Sodium n-Dodecyl Sulfate, Molecular Biology Grade, Dodecyl hydrogen sulfate sodium salt, Sodium n-Dodecyl Sulfate, High Purity
Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 24
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2