
Selamectin

Synonym(s):
(5Z)-25-Cyclohexyl-4′-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-(hydroxyimino)avermectin A1a, Selamectin
Empirical Formula (Hill Notation):
C43H63NO11
CAS Number:
Molecular Weight:
769.96
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing