
Reactive Blue 4

Synonym(s):
1-Amino-4-[3-(4,6-dichlorotriazin-2-ylamino)-4-sulfophenylamino]anthraquinone-2-sulfonic acid, Procion® blue MX-R
Empirical Formula (Hill Notation):
C23H14Cl2N6O8S2
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1