Skip to Content
MilliporeSigma
Propylene glycol British Pharmacopoeia (BP) Reference Standard

Propylene glycol

Propylene glycol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1,2-Propanediol, Propylene glycol
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
EC Number:

Products

Shipping
Product Category
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique