Skip to Content
MilliporeSigma
Propionaldehyde natural, ≥98%, FG

Propionaldehyde

Propionaldehyde natural, ≥98%, FG
Synonym(s):
Propanal, Propionaldehyde
Linear Formula:
CH3CH2CHO
CAS Number:
Molecular Weight:
58.08
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing