Skip to Content
MilliporeSigma
vibrant-m

Powdered milk thistle extract

vibrant-m
Synonym(s):
Silybum marianum extract, Milk thistle extract
CAS Number:

Products

Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1