Skip to Content
MilliporeSigma
Phenobarbital-D5 (ring) solution (deuterium label on ring), 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Phenobarbital-D5 (ring) solution

Phenobarbital-D5 (ring) solution (deuterium label on ring), 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C12D5H7N2O3
CAS Number:
Molecular Weight:
237.27
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique