Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxaloacetic acid
Synonym(s):
2-Oxosuccinic acid, Ketosuccinic acid, Oxalacetic acid, Oxobutanedioic acid
Linear Formula:
HOOCCH
2
COCOOH
CAS Number:
328-42-7
Molecular Weight:
132.07
EC Number:
206-329-8
Products
Product Category
media supplements (1)
substrates (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
O4126
Oxaloacetic acid,
≥97% (HPLC)
Expand
View Pricing
O7753
Oxaloacetic acid,
powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Expand
View Pricing
O9504
Oxaloacetic acid,
powder, suitable for hybridoma
Expand
View Pricing