<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Oryzalin
Empirical Formula (Hill Notation):
C
12
H
18
N
4
O
6
S
CAS Number:
19044-88-3
Molecular Weight:
346.36
EC Number:
242-777-0
Products
Shipping
Ships Today (1)
Product Category
pesticides (1)
Brand
Supelco (1)
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
36182
Oryzalin,
PESTANAL
®
, analytical standard
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More