Skip to Content
MilliporeSigma
Octinoxate United States Pharmacopeia (USP) Reference Standard

Octinoxate

Octinoxate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Ethylhexyl 4-methoxycinnamate, 4-Methoxycinnamic acid 2-ethylhexyl ester, OMC, Octyl methoxycinnamate
Empirical Formula (Hill Notation):
C18H26O3
CAS Number:
Molecular Weight:
290.40

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique