Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Dimethylformamide dimethyl acetal
Synonym(s):
1,1-Dimethoxy-
N
,
N
-dimethylmethylamine, 1,1-Dimethoxytrimethylamine
Linear Formula:
(CH
3
)
2
NCH(OCH
3
)
2
CAS Number:
4637-24-5
Molecular Weight:
119.16
EC Number:
225-063-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technical (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
140732
N
,
N
-Dimethylformamide dimethyl acetal,
technical grade, 94%
Expand
View Pricing
394963
N
,
N
-Dimethylformamide dimethyl acetal,
for GC derivatization, LiChropur
™
Expand
View Pricing