
Methyl Blue

Synonym(s):
Acid blue 93, Aniline blue water soluble, Cotton blue, Poirriers blue, Water blue
Empirical Formula (Hill Notation):
C37H27N3Na2O9S3
Products
Shipping
Product Category
Formula Weight
Physical Form
Agency
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing