Skip to Content
MilliporeSigma
Lithium sulfate monohydrate puriss. p.a., ACS reagent, ≥99.0% (dried basis, T)

Lithium sulfate monohydrate

Lithium sulfate monohydrate puriss. p.a., ACS reagent, ≥99.0% (dried basis, T)
Linear Formula:
Li2SO4 · H2O
CAS Number:
Molecular Weight:
127.96
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing