Skip to Content
MilliporeSigma
(-)-Fenchone phyproof® Reference Substance

L-Fenchone

(-)-Fenchone phyproof® Reference Substance
Synonym(s):
(1R)-(−)-Fenchone, (−)-1,3,3-Trimethyl-2-norbornanone, (−)-Fenchone, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing