Skip to Content
MilliporeSigma
L-Ascorbic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Ascorbic acid

L-Ascorbic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
L-Threoascorbic acid, Antiscorbutic factor, Vitamin C
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Technique