Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kinetin
Synonym(s):
6-Furfurylaminopurine, N
6
-Furfuryladenine
Empirical Formula (Hill Notation):
C
10
H
9
N
5
O
CAS Number:
525-79-1
Molecular Weight:
215.21
EC Number:
208-382-2
Products
Shipping
Ships Today (3)
Product Category
hormones (1)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technical (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
K3378
Kinetin,
suitable for plant cell culture, BioReagent, amorphous powder
Expand
View Pricing
48130
Kinetin,
≥99.0% (HPLC)
Expand
View Pricing
K0753
Kinetin,
suitable for plant cell culture, crystalline
Expand
View Pricing