Isopropyl acetate
Synonym(s):
Acetic acid isopropyl ester, Isopropyl acetate
Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
Products
Shipping
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing