Hydrochloric acid solution
Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Products
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Package Size
Reaction Type
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing