Hyaluronic acid sodium salt from Streptococcus equi
Synonym(s):
Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating, Sodium hyaluronate
CAS Number:
Products
Product Category
Brand
Color
Physical Form
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2