
Histamine dihydrochloride

Synonym(s):
2-(4-Imidazolyl)ethylamine dihydrochloride
Empirical Formula (Hill Notation):
C5H9N3 · 2HCl
CAS Number:
Molecular Weight:
184.07
Products
Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing