
Hexythiazox

Empirical Formula (Hill Notation):
C17H21ClN2O2S
CAS Number:
Molecular Weight:
352.88
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2