Skip to Content
MilliporeSigma
Glycolaldehyde dimer crystalline, mixture of stereoisomers. Melts between 80 and 90 °C depending on stereoisomeric composition

Glycolaldehyde dimer

Glycolaldehyde dimer crystalline, mixture of stereoisomers. Melts between 80 and 90 °C depending on stereoisomeric composition
Synonym(s):
1,4-Dioxane-2,5-diol, 2,5-Dihydroxy-1,4-dioxane, Hydroxyacetaldehyde dimer
Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group