
Dipropylene glycol

Synonym(s):
Bis(hydroxypropyl) ether, Dipropylene glycol
Linear Formula:
HOC3H6OC3H6OH
CAS Number:
Molecular Weight:
134.17
Products
Shipping
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing