Skip to Content
MilliporeSigma
Diphenylmethane 99%

Diphenylmethane

Diphenylmethane 99%
Synonym(s):
Benzylbenzene, Methylenedibenzene, Diphenylmethane
Linear Formula:
(C6H5)2CH2
CAS Number:
Molecular Weight:
168.23

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing