Skip to Content
MilliporeSigma
Diethyl succinate natural, ≥99%, FG

Diethyl succinate

Diethyl succinate natural, ≥99%, FG
Synonym(s):
Diethyl succinate
Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing