Skip to Content
MilliporeSigma
Dibromomethane 99%

Dibromomethane

Dibromomethane 99%
Synonym(s):
Methylene bromide, Dibromomethane
Empirical Formula (Hill Notation):
CH2Br2
CAS Number:
Molecular Weight:
173.83

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing