
Copper(II) sulfate pentahydrate

Synonym(s):
Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate, Copper(II) sulfate pentahydrate
Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
Products
Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Special Grade
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing