Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Cesium chloride
Synonym(s):
Cesium chloride, OmniPur
®
Cesium Chloride
Linear Formula:
CsCl
CAS Number:
7647-17-8
Molecular Weight:
168.36
EC Number:
231-600-2
Products
Shipping
Ships Today (3)
Product Category
salts (12)
density gradient media (3)
Brand
Millipore (1)
Sigma-Aldrich (4)
Supelco (3)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
289329
Cesium chloride,
ReagentPlus
®
, 99.9%
Expand
View Pricing
746487
Cesium chloride,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, 99.9%
Expand
View Pricing
C4036
Cesium chloride,
Molecular Biology, ≥98%
Expand
View Pricing
1.02038
Cesium chloride,
for analysis EMSURE
®
Expand
View Pricing
203025
Cesium chloride,
≥99.999% trace metals basis
Expand
View Pricing
C3032
Cesium chloride,
Molecular Biology, ≥99% (silver nitrate titration)
Expand
View Pricing
C3011
Cesium chloride,
Grade I, ≥99.0%
Expand
View Pricing
31807
Cesium chloride,
puriss. p.a., ≥99.5%
Expand
View Pricing
562599
Cesium chloride,
99.99% trace metals basis
Expand
View Pricing
C3139
Cesium chloride,
optical grade, ≥99.5% trace metals basis
Expand
View Pricing
1.02039
Cesium chloride,
99.995 Suprapur
®
Expand
View Pricing
20966
Cesium chloride,
BioUltra, Molecular Biology, ≥99.5% (AT)
Expand
View Pricing
3030-OP
Cesium chloride,
OmniPur
®
Grade, >= 99.5%
Expand
View Pricing
1.02041
Cesium chloride,
EMPLURA
®
Expand
View Pricing
C6914
Cesium chloride,
Grade II, ≥98%
Expand
View Pricing
940127
Cesium chloride,
≥99.5 (titration), ≥99.9 trace metals basis
Expand
View Pricing
914545
Cesium chloride,
AnhydroBeads
™
, 99.999% trace metals basis, (Perovskite grade)
Expand
View Pricing
C3309
Cesium chloride,
BioXtra, ≥99.5% (titration)
Expand
View Pricing
449733
Cesium chloride,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More