Skip to Content
MilliporeSigma
Cephradine British Pharmacopoeia (BP) Reference Standard

Cephradine

Cephradine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Cefradin
Empirical Formula (Hill Notation):
C16H19N3O4S
CAS Number:
Molecular Weight:
349.40

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing