Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Cellulose
Synonym(s):
Cellulose powder, Cotton linters
CAS Number:
9004-34-6
EC Number:
232-674-9
Products
Shipping
Ships Today (4)
Product Category
carbohydrates (4)
adsorbents (2)
polysaccharides (1)
Brand
Sigma-Aldrich (4)
Supelco (2)
Color
pH Value
Physical Form
Quality Segment
Particle Size (µm)
Polymer Composition
Polymer Type
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
310697
Cellulose,
microcrystalline, powder, 20 μm
Expand
View Pricing
435236
Cellulose,
microcrystalline, powder
Expand
View Pricing
C6288
Cellulose,
fibers, (medium)
Expand
View Pricing
435244
Cellulose,
colloidal, microcrystalline
Expand
View Pricing
22197
Cellulose,
DS-0, powder, suitable for thin layer chromatography (TLC)
Expand
View Pricing
09906
Cellulose,
DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More