Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Carbon tetrachloride
Synonym(s):
Tetrachloromethane
Empirical Formula (Hill Notation):
CCl
4
CAS Number:
56-23-5
Molecular Weight:
153.82
EC Number:
200-262-8
Products
Shipping
Ships Today (3)
Product Category
solvents (2)
pesticides (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
319961
Carbon tetrachloride,
reagent grade, 99.9%
Expand
View Pricing
270652
Carbon tetrachloride,
suitable for HPLC, ≥99.9%
Expand
View Pricing
289116
Carbon tetrachloride,
anhydrous, ≥99.5%
Expand
View Pricing
02671
Carbon tetrachloride,
analytical standard
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More